Learn More
4-Vinylpyridine, 95%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140911000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Vinylpyridine | |
100-43-6,123-31-9 | |
100.0 | |
0.9890g/mL | |
48°C | |
94% min. (GC) | |
C7H7N | |
(C5H4N)CH=Ch2 | |
100 mL | |
0.989 | |
Solubility in water: 29g/L (20°C). Other solubilities: soluble in ethanol,chloroform,in diluted acids,slightly soluble in diethyl ether | |
C=CC1=CC=NC=C1 | |
105.14 | |
7502 | |
105.14 | |
Liquid |
95% | |
94.0 | |
-20°C | |
171°C | |
Authentic | |
Glass bottle | |
1.5500 to 1.5520 | |
MFCD00006447 | |
20, II, 170 | |
4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine | |
KFDVPJUYSDEJTH-UHFFFAOYSA-N | |
4-ethenylpyridine | |
80 to 150ppm hydroquinone | |
<3 cps (20°C) | |
95% |
Chemical Identifiers
100-43-6 | |
105.14 | |
KFDVPJUYSDEJTH-UHFFFAOYSA-N | |
7502 | |
C=CC1=CC=NC=C1 |
C7H7N | |
MFCD00006447 | |
4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine | |
4-ethenylpyridine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life w
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
EINECSNumber : 202-852-0
RTECSNumber : UU1045000
TSCA : TSCA
Recommended Storage : Product may darken during storage
RUO – Research Use Only