missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Vinylpyridine, Stabilized 95%, ACROS Organics™

$53.47 - $104.66

Chemical Identifiers

CAS 100-43-6,123-31-9
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H7N
Linear Formula (C5H4N)CH=Ch2
Formula Weight 105.14
MDL Number MFCD00006447
Chemical Name or Material 4-Vinylpyridine, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC140910010 SDS Thermo Fisher Scientific
AC140910010
Glass bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC140911000 SDS Thermo Fisher Scientific
AC140911000
Glass bottle 100mL Each for $53.47
AC140912500 SDS Thermo Fisher Scientific
140912500
Glass bottle 250mL Each for $104.66
Specifications Description & Specifications

Chemical Identifiers

100-43-6,123-31-9
100.0
C7H7N
105.14
4-Vinylpyridine, 95%
94.0
95%
(C5H4N)CH=Ch2
MFCD00006447
≥94%

Specifications

Liquid
1L
20, II, 170
94.0
Authentic
1.5490 to 1.5520 (20°C, 589nm)
80 to 150ppm hydroquinone
(C5H4N)CH=Ch2
MFCD00006447
4-Vinylpyridine, 95%
105.14
Clear colorless to yellow-brown
0.9890g/mL
100.0
Product may darken during storage
123-31-9,100-43-6
Glass bottle
C7H7N
95%
≥94%

Safety and Handling

GHS H Statement
Flammable liquid and vapor. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 202-852-0

RTECSNumber : UU1045000

TSCA : TSCA