Please login to your online account to display your discounted pricing

4-Vinylphenol, 95%, 10% solution in propylene glycol, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC434041000

 View more versions of this product

Catalog No. AC434041000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 120.16
Physical Form Liquid
Quantity 100mL
Color Clear colorless to orange
Density 1.039g/mL
CAS Max % 11.0
CAS Min % 9.0
CAS Max % 91.0
CAS Min % 89.0
Synonym 4-Ethenylphenol
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Refractive Index 1.4420 to 1.4520 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H8O
CAS 57-55-6
Chemical Name or Material 4-Vinylphenol
Assay Percent Range 95%
CAS 2628-17-3
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Vinylphenol
CAS 2628-17-3
CAS Min % 89.0
CAS Max % 91.0
CAS 57-55-6
CAS Min % 9.0
CAS Max % 11.0
Assay Percent Range 95%
Molecular Formula C8H8O
Formula Weight 120.16
Synonym 4-Ethenylphenol
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.