missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Vinylphenol, 95%, 10% solution in propylene glycol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 434040250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Vinylphenol | |
89.0,9.0 | |
Colorless to Orange | |
189°C | |
9 to 15% (GC) | |
C8H8O | |
25 mL | |
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene | |
C=CC1=CC=C(C=C1)O | |
120.16 | |
CHEBI:1883 | |
95% |
2628-17-3,57-55-6 | |
91.0,11.0 | |
1.0390g/mL | |
Authentic | |
Glass bottle | |
1.4420 to 1.4520 (20°C, 589nm) | |
1.039 | |
FUGYGGDSWSUORM-UHFFFAOYSA-N | |
4-ethenylphenol | |
62453 | |
120.16 | |
Liquid |
Chemical Identifiers
2628-17-3 | |
120.16 | |
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene | |
CHEBI:1883 | |
C=CC1=CC=C(C=C1)O |
C8H8O | |
FUGYGGDSWSUORM-UHFFFAOYSA-N | |
62453 | |
4-ethenylphenol |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 220-103-6
RUO – Research Use Only