missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Vinylbenzoic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC294912500

 View more versions of this product

Catalog No. AC294912500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Vinylbenzoic acid
Glass Bottle
Beige to White
250mg
95.0
96%
CH2CHC6H4COOH
4-carboxystyrene, 4-vinyl benzoic acid, 4-vinyl-benzoic acid, 4-vinylbenzoic acid, benzoic acid, 4-ethenyl, p-vinyl benzoic acid, p-vinylbenzoic acid, para-vinyl benzoic acid, pubchem12624, styrene-4-carboxylic acid
IRQWEODKXLDORP-UHFFFAOYSA-N
4-ethenylbenzoic acid
14098
Powder
Authentic
0.1% t-butylcatechol
142.0°C to 144.0°C
1075-49-6
100.0
C9H8O2
MFCD00002569
(10% in Methanol) Clear solution
C=CC1=CC=C(C=C1)C(=O)O
148.16
148.16
96%

Chemical Identifiers

1075-49-6
148.16
IRQWEODKXLDORP-UHFFFAOYSA-N
14098
C=CC1=CC=C(C=C1)C(=O)O
C9H8O2
MFCD00002569
4-carboxystyrene, 4-vinyl benzoic acid, 4-vinyl-benzoic acid, 4-vinylbenzoic acid, benzoic acid, 4-ethenyl, p-vinyl benzoic acid, p-vinylbenzoic acid, para-vinyl benzoic acid, pubchem12624, styrene-4-carboxylic acid
4-ethenylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-053-4

SDS Documents