Learn More
4-Vinylaniline, 90%, technical, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 308480010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Vinylaniline | |
88.0 | |
1.0100g/mL | |
99°C | |
88% min. (GC) | |
C8H9N | |
H2C=CHC6H4NH2 | |
1g | |
4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene | |
C=CC1=CC=C(C=C1)N | |
119.17 | |
119.17 | |
Technical |
1520-21-4 | |
100.0 | |
249°C | |
Authentic | |
Glass bottle | |
1.6250 to 1.629 | |
MFCD00015329 | |
1.01 | |
LBSXSAXOLABXMF-UHFFFAOYSA-N | |
4-ethenylaniline | |
73700 | |
90% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Suspected of causing cancer
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Danger
EINECSNumber : 216-185-8
RUO â Research Use Only