Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Undecanone, 98%

$42.47 - $415.61

Chemical Identifiers

Chemical Name or Material 4-Undecanone
CAS 14476-37-0
Molecular Formula C11H22O
Formula Weight 170.3
MDL Number MFCD00048923
Beilstein 1749499
Synonym n-Heptyl n-propyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2013506

  SDS
alfa aesar™
B2013506
5g Each for $42.47

AAB2013522

  SDS
alfa aesar™
B2013522
100g Each for $415.61
Description & Specifications

Specifications

Formula Weight 170.3
Chemical Name or Material 4-Undecanone
Assay 98%
Molecular Formula C11H22O
MDL Number MFCD00048923
CAS 14476-37-0
Beilstein 1749499
Synonym n-Heptyl n-propyl ketone
Boiling Point 104° to 107°C (13mmHg)
Quantity 5g
Flash Point 92°C (197°F)