missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Tritylphenol, 98%, Thermo Scientific Chemicals
$69.79 - $248.75
Chemical Identifiers
CAS | 978-86-9 |
---|---|
Molecular Formula | C25H20O |
Molecular Weight (g/mol) | 336.43 |
MDL Number | MFCD00002364 |
InChI Key | NIPKXTKKYSKEON-UHFFFAOYSA-N |
Synonym | 4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl |
PubChem CID | 70422 |
IUPAC Name | 4-tritylphenol |
SMILES | OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0607306
|
Thermo Scientific Chemicals
L0607306 |
5 g |
Each for $69.79
|
|
|||||
AAL0607314
|
Thermo Scientific Chemicals
L0607314 |
25 g |
Each for $248.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
978-86-9 | |
336.43 | |
NIPKXTKKYSKEON-UHFFFAOYSA-N | |
70422 | |
OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C25H20O | |
MFCD00002364 | |
4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl | |
4-tritylphenol |
Specifications
978-86-9 | |
C25H20O | |
5 g | |
4-triphenylmethylphenol, 4-triphenylmethyl phenol, p-tritylphenol, p-triphenylmethylphenol, phenol, 4-triphenylmethyl, p-cresol, .alpha.,.alpha.,.alpha.-triphenyl, 4-tritylphenol, p-triphenylmethyl phenol, 4-tri phenyl methyl phenol, phenol,4-triphenylmethyl | |
OC1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
336.43 | |
336.43 | |
4-Tritylphenol |
276°C to 286°C | |
MFCD00002364 | |
2290531 | |
NIPKXTKKYSKEON-UHFFFAOYSA-N | |
4-tritylphenol | |
70422 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-557-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only