missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trimethylsilyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Manufacturer:  TCI America T26645G

 View more versions of this product

Catalog No. T26645G



Specifications

Specifications

4-(Trimethylsilyl)phenylboronic Acid (contains varying amounts of Anhydride)
5g
C9H15BO2Si
4-(Trimethylsilyl)benzeneboronic Acid
B(C1=CC=C(C=C1)[Si](C)(C)C)(O)O
194.112
194.11
White
17865-11-1
MFCD00093348
NRPZMSUGPMYBCQ-UHFFFAOYSA-N
(4-trimethylsilylphenyl)boronic acid
1710
Crystal-Powder at 20°C

Chemical Identifiers

17865-11-1
194.112
NRPZMSUGPMYBCQ-UHFFFAOYSA-N
1710
B(C1=CC=C(C=C1)[Si](C)(C)C)(O)O
C9H15BO2Si
MFCD00093348
4-(Trimethylsilyl)benzeneboronic Acid
(4-trimethylsilylphenyl)boronic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents