Learn More
4-(Trimethylsilyl)morpholine, 97%, Thermo Scientific Chemicals
$108.48 - $511.33
Chemical Identifiers
CAS | 13368-42-8 |
---|---|
Molecular Formula | C7H17NOSi |
Molecular Weight (g/mol) | 159.304 |
MDL Number | MFCD00010098 |
InChI Key | JJOWIQMPCCUIGA-UHFFFAOYSA-N |
Synonym | 4-trimethylsilyl morpholine, n-trimethylsilyl morpholine, n-trimethylsilylmorpholine, trimethyl morpholin-4-yl silane, morpholine, 4-trimethylsilyl, trimethylsilyl morpholin, morpholinotrimethylsilane, trimethylmorpholinosilane, acmc-1btin, timtec-bb sbb009054 |
PubChem CID | 559763 |
IUPAC Name | trimethyl(morpholin-4-yl)silane |
SMILES | C[Si](C)(C)N1CCOCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5522906
|
Thermo Scientific Chemicals
H5522906 |
5 g |
Each for $108.48
|
|
|||||
AAH5522914
|
Thermo Scientific Chemicals
H5522914 |
25 g |
Each for $511.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13368-42-8 | |
159.304 | |
JJOWIQMPCCUIGA-UHFFFAOYSA-N | |
559763 | |
C[Si](C)(C)N1CCOCC1 |
C7H17NOSi | |
MFCD00010098 | |
4-trimethylsilyl morpholine, n-trimethylsilyl morpholine, n-trimethylsilylmorpholine, trimethyl morpholin-4-yl silane, morpholine, 4-trimethylsilyl, trimethylsilyl morpholin, morpholinotrimethylsilane, trimethylmorpholinosilane, acmc-1btin, timtec-bb sbb009054 | |
trimethyl(morpholin-4-yl)silane |
Specifications
13368-42-8 | |
160°C | |
C7H17NOSi | |
MFCD00010098 | |
UN2924 | |
4-trimethylsilyl morpholine, n-trimethylsilyl morpholine, n-trimethylsilylmorpholine, trimethyl morpholin-4-yl silane, morpholine, 4-trimethylsilyl, trimethylsilyl morpholin, morpholinotrimethylsilane, trimethylmorpholinosilane, acmc-1btin, timtec-bb sbb009054 | |
C[Si](C)(C)N1CCOCC1 | |
159.304 | |
159.3 | |
4-(Trimethylsilyl)morpholine |
0.897 | |
29°C (84°F) | |
1.441 | |
5 g | |
1071480 | |
JJOWIQMPCCUIGA-UHFFFAOYSA-N | |
trimethyl(morpholin-4-yl)silane | |
559763 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Hazard Class: 3; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only