Please login to your online account to display your discounted pricing

4-(Trimethylsilyl)morpholine, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369200050

 View more versions of this product

Catalog No. AC369200050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Formula Weight 159.3
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
Density 0.897g/mL
CAS Max % 100.0
CAS Min % 95.0
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Infrared Spectrum Authentic
CAS 13368-42-8
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C7H17NOSi
MDL Number MFCD00010098
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(Trimethylsilyl)morpholine

Specifications

Chemical Name or Material 4-(Trimethylsilyl)morpholine
CAS 13368-42-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H17NOSi
Formula Weight 159.3
MDL Number MFCD00010098
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

SDS & Safety Documents