Please login to your online account to display your discounted pricing

4-(Trimethylsilyl)morpholine, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Trimethylsilyl)morpholine
CAS 13368-42-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H17NOSi
Formula Weight 159.3
MDL Number MFCD00010098
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369200050

  SDS
thermo fisher scientific
AC369200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 159.3
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
Density 0.897g/mL
CAS Max % 100.0
CAS Min % 95.0
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Infrared Spectrum Authentic
CAS 13368-42-8
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C7H17NOSi
MDL Number MFCD00010098
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(Trimethylsilyl)morpholine
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.