Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester, 97%

$118.68 - $467.95

Chemical Identifiers

Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
CAS 870238-65-6
Molecular Formula C17H25BO2Si
Formula Weight 300.28
MDL Number MFCD08706286
Synonym 4,4,5,5-Tetramethyl-2-(4-[(trimethylsilyl)ethynyl]phenyl)-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5169703

 
alfa aesar™
H5169703
1g Each for $118.68

AAH5169706

 
alfa aesar™
H5169706
5g Each for $467.95
Description & Specifications

Specifications

Formula Weight 300.28
Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C17H25BO2Si
MDL Number MFCD08706286
CAS 870238-65-6
Synonym 4,4,5,5-Tetramethyl-2-(4-[(trimethylsilyl)ethynyl]phenyl)-1,3,2-dioxaborolane
Quantity 1g