missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5506014

 View more versions of this product

Catalog No. AAH5506014



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at 4°C. Keep away from oxidizing agents.
Specifications

Specifications

4-[(Trimethylsilyl)ethynyl]benzaldehyde
25g
C12H14OSi
4-2-trimethylsilyl ethynyl benzaldehyde
C[Si](C)(C)C#CC1=CC=C(C=C1)C=O
202.328
202.33
66°C to 70°C
77123-57-0
MFCD02093765
UZQDUXAJFTWMDT-UHFFFAOYSA-N
4-(2-trimethylsilylethynyl)benzaldehyde
2771643
97%

Chemical Identifiers

77123-57-0
202.328
UZQDUXAJFTWMDT-UHFFFAOYSA-N
2771643
C[Si](C)(C)C#CC1=CC=C(C=C1)C=O
C12H14OSi
MFCD02093765
4-2-trimethylsilyl ethynyl benzaldehyde
4-(2-trimethylsilylethynyl)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents