missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trimethylsiloxy)benzaldehyde, 97%

$33.51 - $109.34

Chemical Identifiers

CAS 1012-12-0
Molecular Formula C10H14O2Si
Formula Weight 194.31
MDL Number MFCD00040723
Beilstein 2085051
Chemical Name or Material 4-(Trimethylsiloxy)benzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1346303 SDS Alfa Aesar™
L1346303
1g Each for $33.51
AAL1346306 SDS Alfa Aesar™
L1346306
5g Each for $109.34
Specifications Description & Specifications

Chemical Identifiers

1012-12-0
194.31
2085051
C10H14O2Si
MFCD00040723
4-(Trimethylsiloxy)benzaldehyde

Specifications

83° to 84°C (3mmHg)
1012-12-0
194.31
2085051
97%
1g
C10H14O2Si
MFCD00040723
4-(Trimethylsiloxy)benzaldehyde

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning