missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)phenylacetic acid, 98%

Manufacturer:  Alfa Aesar™ H63432MD

 View more versions of this product

Catalog No. AAH63432MD



Specifications

Specifications

4-(Trifluoromethylthio)phenylacetic acid
243977-23-3
MFCD00236351
BFDKCZMYQOSTJG-UHFFFAOYSA-N
2-[4-(trifluoromethylsulfanyl)phenyl]acetic acid
2777902
Solid
250mg
C9H7F3O2S
2-4-trifluoromethyl thio phenyl acetic acid, 2-4-trifluoromethylthio phenyl acetic acid, 4-trifluoromethyl sulfanyl phenyl acetic acid, 4-trifluoromethylsulfanyl-phenyl-acetic acid, 4-trifluoromethylthio phenylacetic acid, acmc-1chv0, benzeneacetic acid, 4-trifluoromethyl thio, pubchem7296
C1=CC(=CC=C1CC(=O)O)SC(F)(F)F
236.208
236.21
98%

Chemical Identifiers

243977-23-3
236.208
BFDKCZMYQOSTJG-UHFFFAOYSA-N
2777902
C1=CC(=CC=C1CC(=O)O)SC(F)(F)F
C9H7F3O2S
MFCD00236351
2-4-trifluoromethyl thio phenyl acetic acid, 2-4-trifluoromethylthio phenyl acetic acid, 4-trifluoromethyl sulfanyl phenyl acetic acid, 4-trifluoromethylsulfanyl-phenyl-acetic acid, 4-trifluoromethylthio phenylacetic acid, acmc-1chv0, benzeneacetic acid, 4-trifluoromethyl thio, pubchem7296
2-[4-(trifluoromethylsulfanyl)phenyl]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents