Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)phenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)phenyl isocyanate
CAS 24032-84-6
Molecular Formula C8H4F3NOS
Formula Weight 219.19
MDL Number MFCD00036025
UN Number UN2206
Beilstein 6290747
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1920603

  SDS
alfa aesar™
L1920603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.19
Assay 97%
CAS 24032-84-6
Beilstein 6290747
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Quantity 1g
Chemical Name or Material 4-(Trifluoromethylthio)phenyl isocyanate
MDL Number MFCD00036025
Health Hazard 2 GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 175°C
Molecular Formula C8H4F3NOS
UN Number UN2206
Flash Point 80°C (176°F)

Solubility
It hydrolyzes with water.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C