Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)phenol, 98%

$87.70 - $334.50

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)phenol
CAS 461-84-7
Molecular Formula C7H5F3OS
Formula Weight 194.18
MDL Number MFCD00036035
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2499603

  SDS
alfa aesar™
B2499603
1g Each for $87.70

AAB2499606

  SDS
alfa aesar™
B2499606
5g Each for $334.50
Description & Specifications

Specifications

Formula Weight 194.18
Chemical Name or Material 4-(Trifluoromethylthio)phenol
Assay 98%
Molecular Formula C7H5F3OS
MDL Number MFCD00036035
CAS 461-84-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 77° to 78°C (7mmHg)
Melting Point 57° to 60°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.