Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)benzylamine, 95%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)benzylamine
CAS 128273-56-3
Molecular Formula C8H8F3NS
Formula Weight 207.21
MDL Number MFCD00190129
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3151503

  SDS
alfa aesar™
H3151503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3151506

  SDS
alfa aesar™
H3151506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.21
UN Number UN2735
Chemical Name or Material 4-(Trifluoromethylthio)benzylamine
Quantity 1g
Boiling Point 224° to 225°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
CAS 128273-56-3
MDL Number MFCD00190129
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C8H8F3NS
Flash Point 80°C (176°F)
Assay 95%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.