Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)benzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)benzyl bromide
CAS 21101-63-3
Molecular Formula C8H6BrF3S
Formula Weight 271.1
MDL Number MFCD00129189
UN Number UN3261
Beilstein 2209970
Synonym alpha-Bromo-4-(trifluoromethylthio)toluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1920503

  SDS
alfa aesar™
L1920503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1920506

  SDS
alfa aesar™
L1920506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19205MD

  SDS
alfa aesar™
L19205MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.1
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 4-(Trifluoromethylthio)benzyl bromide
Molecular Formula C8H6BrF3S
Odor Pungent
Assay 97%
MDL Number MFCD00129189
CAS 21101-63-3
UN Number UN3261
Beilstein 2209970
Synonym alpha-Bromo-4-(trifluoromethylthio)toluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 115° to 118°C (13mmHg)
Melting Point 53° to 56°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.