Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)benzyl alcohol, 97%

$64.18 - $115.41

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)benzyl alcohol
CAS 56456-52-1
Molecular Formula C8H7F3OS
Formula Weight 208.2
MDL Number MFCD00236342
Beilstein 8616292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2402601

  SDS
alfa aesar™
B2402601
0.25g Each for $64.18

AAB2402603

  SDS
alfa aesar™
B2402603
1g Each for $115.41
Description & Specifications

Specifications

Formula Weight 208.2
Molecular Formula C8H7F3OS
Melting Point 49° to 54°C
MDL Number MFCD00236342
Beilstein 8616292
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 0.25g
Chemical Name or Material 4-(Trifluoromethylthio)benzyl alcohol
Health Hazard 1 Warning
Assay 97%
CAS 56456-52-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.