missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Trifluoromethylthio)benzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America T27455G
Specifications
4-(Trifluoromethylthio)benzoyl Chloride | |
Yellow | |
C8H4ClF3OS | |
5 g | |
4-trifluoromethylthio benzoyl chloride, 4-trifluoromethyl thio benzoyl chloride, 4-trifluoromethylsulfanyl benzoyl chloride, 4-trifluoromethyl sulfanyl benzoyl chloride, 4-trifluomethylthio benzoyl chloride, benzoyl chloride, 4-trifluoromethyl thio, 4-trifluoromethyl sulphanyl benzoyl chloride, 4-trifluoromethylsulfanyl-benzoyl chloride, acmc-209hxt, 4-trifluoromethylthiobenzoylchloride | |
C1=CC(=CC=C1C(=O)Cl)SC(F)(F)F | |
240.624 | |
240.62 | |
Liquid |
330-14-3 | |
230°C | |
MFCD01631632 | |
2920 | |
BCMFTOCCLOAGBP-UHFFFAOYSA-N | |
4-(trifluoromethylsulfanyl)benzoyl chloride | |
2777865 | |
≥98.0% (GC,T) |
Chemical Identifiers
330-14-3 | |
240.624 | |
BCMFTOCCLOAGBP-UHFFFAOYSA-N | |
2777865 | |
C1=CC(=CC=C1C(=O)Cl)SC(F)(F)F |
C8H4ClF3OS | |
MFCD01631632 | |
4-trifluoromethylthio benzoyl chloride, 4-trifluoromethyl thio benzoyl chloride, 4-trifluoromethylsulfanyl benzoyl chloride, 4-trifluoromethyl sulfanyl benzoyl chloride, 4-trifluomethylthio benzoyl chloride, benzoyl chloride, 4-trifluoromethyl thio, 4-trifluoromethyl sulphanyl benzoyl chloride, 4-trifluoromethylsulfanyl-benzoyl chloride, acmc-209hxt, 4-trifluoromethylthiobenzoylchloride | |
4-(trifluoromethylsulfanyl)benzoyl chloride |
Safety and Handling
TSCA : No