missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethylthio)benzoyl Chloride, TCI America™

Manufacturer:  TCI America T274525G

 View more versions of this product

Catalog No. T274525G



Specifications

Specifications

4-(Trifluoromethylthio)benzoyl Chloride
Yellow
330-14-3
MFCD01631632
4-trifluoromethylthio benzoyl chloride, 4-trifluoromethyl thio benzoyl chloride, 4-trifluoromethylsulfanyl benzoyl chloride, 4-trifluoromethyl sulfanyl benzoyl chloride, 4-trifluomethylthio benzoyl chloride, benzoyl chloride, 4-trifluoromethyl thio, 4-trifluoromethyl sulphanyl benzoyl chloride, 4-trifluoromethylsulfanyl-benzoyl chloride, acmc-209hxt, 4-trifluoromethylthiobenzoylchloride
C1=CC(=CC=C1C(=O)Cl)SC(F)(F)F
240.624
240.62
≥98.0% (GC,T)
230°C
25g
C8H4ClF3OS
2920
BCMFTOCCLOAGBP-UHFFFAOYSA-N
4-(trifluoromethylsulfanyl)benzoyl chloride
2777865
Clear Liquid at 20°C

Chemical Identifiers

330-14-3
240.624
BCMFTOCCLOAGBP-UHFFFAOYSA-N
2777865
C1=CC(=CC=C1C(=O)Cl)SC(F)(F)F
C8H4ClF3OS
MFCD01631632
4-trifluoromethylthio benzoyl chloride, 4-trifluoromethyl thio benzoyl chloride, 4-trifluoromethylsulfanyl benzoyl chloride, 4-trifluoromethyl sulfanyl benzoyl chloride, 4-trifluomethylthio benzoyl chloride, benzoyl chloride, 4-trifluoromethyl thio, 4-trifluoromethyl sulphanyl benzoyl chloride, 4-trifluoromethylsulfanyl-benzoyl chloride, acmc-209hxt, 4-trifluoromethylthiobenzoylchloride
4-(trifluoromethylsulfanyl)benzoyl chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents