Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethylthio)benzoic acid, 97%

$825.70 - $825.70

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)benzoic acid
CAS 330-17-6
Molecular Formula C8H5F3O2S
Formula Weight 222.18
MDL Number MFCD00040906
Beilstein 2693449
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2010414

  SDS
alfa aesar™
B2010414
25g Each for $825.70
Description & Specifications

Specifications

Formula Weight 222.18
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Chemical Name or Material 4-(Trifluoromethylthio)benzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00040906
CAS 330-17-6
Assay 97%
Melting Point 159° to 162°C
Molecular Formula C8H5F3O2S
Beilstein 2693449
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.