missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethylthio)aniline, TCI America™

Manufacturer:  TCI America T277525G

 View more versions of this product

Catalog No. T277525G



Specifications

Specifications

4-(Trifluoromethylthio)aniline
Yellow
372-16-7
MFCD00040926
OHHHTUXVBNGOGI-UHFFFAOYSA-N
4-(trifluoromethylsulfanyl)aniline
123054
Clear Liquid at 20°C
103°C
25g
C7H6F3NS
4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
C1=CC(=CC=C1N)SC(F)(F)F
193.187
193.19
≥98.0% (GC,T)

Chemical Identifiers

372-16-7
193.187
OHHHTUXVBNGOGI-UHFFFAOYSA-N
123054
C1=CC(=CC=C1N)SC(F)(F)F
C7H6F3NS
MFCD00040926
4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, 4-trifluoromethyl sulfanyl aniline, 4-aminophenyl trifluoromethyl sulfide, 4-aminophenyl trifluoromethyl sulphide, 4-trifluoromethylsulfanyl aniline, 4-trifluoromethylthio aniline, 4-trifluoromethyl thio aniline, benzenamine, 4-trifluoromethyl thio, 4-trifluoromethylthio phenylamine
4-(trifluoromethylsulfanyl)aniline
Safety and Handling

Safety and Handling

TSCA : No

Documents