Please login to your online account to display your discounted pricing

4-(Trifluoromethylthio)aniline, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethylthio)aniline
CAS 372-16-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H6F3NS
Formula Weight 193.19
MDL Number MFCD00040926
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC371940050

  SDS
thermo fisher scientific
AC371940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC371940010

  SDS
thermo fisher scientific
AC371940010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 193.19
Quantity 5g
Color Clear colorless to yellow
Density 1.3510g/mL
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-(Trifluoromethylthio)aniline
Assay Percent Range 95%
CAS 372-16-7
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H6F3NS
MDL Number MFCD00040926
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.