Please login to your online account to display your discounted pricing

4-(Trifluoromethylthio)aniline, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371940010

 View more versions of this product

Catalog No. AC371940010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 193.19
Assay ≥94%
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow
Density 1.3510g/mL
CAS Max % 100.0
CAS Min % 94.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 372-16-7
Chemical Name or Material 4-(Trifluoromethylthio)aniline
Assay Percent Range 95%
Molecular Formula C7H6F3NS
MDL Number MFCD00040926
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)

Specifications

Chemical Name or Material 4-(Trifluoromethylthio)aniline
CAS 372-16-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H6F3NS
Formula Weight 193.19
MDL Number MFCD00040926
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents