Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Trifluoromethylthio)acetophenone, 97%

$103.14 - $307.85

Chemical Identifiers

Chemical Name or Material 4'-(Trifluoromethylthio)acetophenone
CAS 713-67-7
Molecular Formula C9H7F3OS
Formula Weight 220.21
MDL Number MFCD00236332
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2660503

 
alfa aesar™
H2660503
1g Each for $103.14

AAH2660506

 
alfa aesar™
H2660506
5g Each for $307.85
Description & Specifications

Specifications

Formula Weight 220.21
Chemical Name or Material 4'-(Trifluoromethylthio)acetophenone
Assay 97%
Molecular Formula C9H7F3OS
MDL Number MFCD00236332
CAS 713-67-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 28° to 32°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.