missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)thiobenzamide, TCI America™

Manufacturer:  TCI America T277125G

 View more versions of this product

Catalog No. T277125G



Specifications

Specifications

4-(Trifluoromethyl)thiobenzamide
137°C
72505-21-6
MFCD00051806
4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl thiobenzamine, 4-trifluoromethyl thiobenzamide
C1=CC(=CC=C1C(=S)N)C(F)(F)F
205.198
205.20
≥98.0% (HPLC)
Undesignated
25g
C8H6F3NS
2811
IPRFNMJROWWFBH-UHFFFAOYSA-N
4-(trifluoromethyl)benzenecarbothioamide
2734823
Crystal-Powder at 20°C

Chemical Identifiers

72505-21-6
205.198
IPRFNMJROWWFBH-UHFFFAOYSA-N
2734823
C1=CC(=CC=C1C(=S)N)C(F)(F)F
C8H6F3NS
MFCD00051806
4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl thiobenzamine, 4-trifluoromethyl thiobenzamide
4-(trifluoromethyl)benzenecarbothioamide
Safety and Handling

Safety and Handling

TSCA : No

Documents