missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)thiobenzamide, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1460003

 View more versions of this product

Catalog No. AAA1460003



Specifications

Specifications

4-(Trifluoromethyl)thiobenzamide
1g
C8H6F3NS
UN2811
4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl thiobenzamine, 4-trifluoromethyl thiobenzamide
C1=CC(=CC=C1C(=S)N)C(F)(F)F
205.198
205.2
135°C to 138°C
72505-21-6
MFCD00051806
4672885
IPRFNMJROWWFBH-UHFFFAOYSA-N
4-(trifluoromethyl)benzenecarbothioamide
2734823
97%

Chemical Identifiers

72505-21-6
205.198
IPRFNMJROWWFBH-UHFFFAOYSA-N
2734823
C1=CC(=CC=C1C(=S)N)C(F)(F)F
C8H6F3NS
MFCD00051806
4-trifluoromethyl thiobenzamide, 4-trifluoromethyl benzenecarbothioamide, 4-trifluoromethylthiobenzamide, 4-trifluoromethyl benzene-1-carbothioamide, 4-trifluoromethyl benzothioamide, 4-trifluoromethyl-thiobenzamide, benzenecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl thiobenzamide, 4-trifluoromethyl thiobenzamine, 4-trifluoromethyl thiobenzamide
4-(trifluoromethyl)benzenecarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents