missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)styrene, 98%, stab.

Manufacturer:  Alfa Aesar™ H5351306

 View more versions of this product

Catalog No. AAH5351306



Description

Description

Applications
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Moisture & light sensitive. Incompatible with oxidizing agents, acids, bases, heat, ultravioloet radiation and free radical initiators.
Specifications

Specifications

4-(Trifluoromethyl)styrene
42°C (108°F)
Light and moisture sensitive
5g
402-50-6
MFCD00075539
1863548
Insoluble in water.
C=CC1=CC=C(C=C1)C(F)(F)F
172.15
172.15
1.165
1.466
65°C to 66°C (40mmHg)
stabilized
C9H7F3
UN1993
1-ethenyl-4-trifluoromethyl benzene
CEWDRCQPGANDRS-UHFFFAOYSA-N
1-ethenyl-4-(trifluoromethyl)benzene
2782798
98%

Chemical Identifiers

402-50-6
172.15
CEWDRCQPGANDRS-UHFFFAOYSA-N
2782798
C=CC1=CC=C(C=C1)C(F)(F)F
C9H7F3
MFCD00075539
1-ethenyl-4-trifluoromethyl benzene
1-ethenyl-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

Documents