missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)quinoline-2-carboxylic acid, 95%

$422.54 - $422.54

Chemical Identifiers

CAS 588702-67-4
Molecular Formula C11H6F3NO2
Molecular Weight (g/mol) 241.169
MDL Number MFCD06796588
InChI Key YGGNZDRZJWUYRO-UHFFFAOYSA-N
Synonym 4-trifluoromethyl quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylicacid, 4-trifluoromethyl-2-quinolinecarboxylic acid, 2-quinolinecarboxylicacid, 4-trifluoromethyl, 4-trifluoromethyl quinoline-2-carboxylic acid
PubChem CID 11085912
IUPAC Name 4-(trifluoromethyl)quinoline-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC(=N2)C(=O)O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6151303 View Documents Alfa Aesar™
H6151303
1g Each for $422.54
 
Specifications

Chemical Identifiers

588702-67-4
241.169
YGGNZDRZJWUYRO-UHFFFAOYSA-N
11085912
C1=CC=C2C(=C1)C(=CC(=N2)C(=O)O)C(F)(F)F
C11H6F3NO2
MFCD06796588
4-trifluoromethyl quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylicacid, 4-trifluoromethyl-2-quinolinecarboxylic acid, 2-quinolinecarboxylicacid, 4-trifluoromethyl, 4-trifluoromethyl quinoline-2-carboxylic acid
4-(trifluoromethyl)quinoline-2-carboxylic acid

Specifications

1g
C11H6F3NO2
4-trifluoromethyl quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylic acid, 4-trifluoromethyl-quinoline-2-carboxylicacid, 4-trifluoromethyl-2-quinolinecarboxylic acid, 2-quinolinecarboxylicacid, 4-trifluoromethyl, 4-trifluoromethyl quinoline-2-carboxylic acid
C1=CC=C2C(=C1)C(=CC(=N2)C(=O)O)C(F)(F)F
241.169
241.17
4-(Trifluoromethyl)quinoline-2-carboxylic acid
588702-67-4
MFCD06796588
YGGNZDRZJWUYRO-UHFFFAOYSA-N
4-(trifluoromethyl)quinoline-2-carboxylic acid
11085912
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C