Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)pyridine-3-carbothioamide, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
CAS 158063-54-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5F3N2S
Formula Weight 206.19
MDL Number MFCD00205806
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05766CB

 
fisher scientific company
SPB05766CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPB05766DA

 
fisher scientific company
SPB05766DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.19
Quantity 250mg
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
Assay Percent Range 95%
MDL Number MFCD00205806
Packaging Amber glass bottle
Molecular Formula C7H5F3N2S
CAS 158063-54-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.