Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)pyridine-3-carbothioamide, ≥95%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SPB05766CB

 View more versions of this product

Catalog No. SPB05766CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.19
Quantity 250mg
Assay 95%
CAS Min % 95.0
Packaging Amber glass bottle
Assay Percent Range 95%
MDL Number MFCD00205806
Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
Molecular Formula C7H5F3N2S
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 158063-54-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
CAS 158063-54-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5F3N2S
Formula Weight 206.19
MDL Number MFCD00205806
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents