Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Trifluoromethyl)propiophenone, 97%

$59.96 - $59.96

Chemical Identifiers

Chemical Name or Material 4'-(Trifluoromethyl)propiophenone
CAS 711-33-1
Molecular Formula C10H9F3O
Formula Weight 202.18
MDL Number MFCD00039231
Beilstein 1874014
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2007403

  SDS
alfa aesar™
B2007403
1g Each for $59.96

AAB2007406

  SDS
alfa aesar™
B2007406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4'-(Trifluoromethyl)propiophenone
Flash Point 98°C (208°F)
Boiling Point 62°C (2mmHg)
Melting Point 35° to 37°C
Quantity 1g
CAS 711-33-1
Assay 97%
Molecular Formula C10H9F3O
Formula Weight 202.18
MDL Number MFCD00039231
Beilstein 1874014