Please login to your online account to display your discounted pricing

Alfa Aesar™ [4-(Trifluoromethyl)phenylthio]acetic acid

$136.81 - $602.70

Chemical Identifiers

Chemical Name or Material [4-(Trifluoromethyl)phenylthio]acetic acid
CAS 102582-93-4
Molecular Formula C9H7F3O2S
Formula Weight 236.21
MDL Number MFCD00236351
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5028306

  SDS
alfa aesar™
H5028306
5g Each for $602.70

AAH5028303

  SDS
alfa aesar™
H5028303
1g Each for $136.81
Description & Specifications

Specifications

Formula Weight 236.21
Molecular Formula C9H7F3O2S
MDL Number MFCD00236351
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
CAS 102582-93-4
Health Hazard 1 Warning
Chemical Name or Material [4-(Trifluoromethyl)phenylthio]acetic acid
Melting Point 114° to 118°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.