Please login to your online account to display your discounted pricing

Alfa Aesar™ [4-(Trifluoromethyl)phenylsulfonyl]acetonitrile, 98%

$108.34 - $268.20

Chemical Identifiers

Chemical Name or Material [4-(Trifluoromethyl)phenylsulfonyl]acetonitrile
CAS 186405-37-8
Molecular Formula C9H6F3NO2S
Formula Weight 249.21
MDL Number MFCD08741474
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2753803

  SDS
alfa aesar™
H2753803
1g Each for $268.20

AAH27538MD

  SDS
alfa aesar™
H27538MD
250mg Each for $108.34
Description & Specifications

Specifications

Formula Weight 249.21
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 186405-37-8
Quantity 1g
Melting Point 142° to 145°C
Chemical Name or Material [4-(Trifluoromethyl)phenylsulfonyl]acetonitrile
Health Hazard 1 Warning
Molecular Formula C9H6F3NO2S
Assay 98%
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD08741474
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.