Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)phenylhydrazine, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
CAS 368-90-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH2
Formula Weight 176.14
MDL Number MFCD00042508
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC397950050

  SDS
thermo fisher scientific
AC397950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC397950010

  SDS
thermo fisher scientific
AC397950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥95%
Formula Weight 176.14
Quantity 5g
Color Yellow to brown
Melting Point 59° to 65°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH2
CAS 368-90-1
Packaging Glass bottle
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
MDL Number MFCD00042508
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.