Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)phenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)phenylacetonitrile
CAS 2338-75-2
Molecular Formula C9H6F3N
Formula Weight 185.15
MDL Number MFCD00001921
UN Number UN3439
Beilstein 909641
Synonym 4-(Trifluoromethyl)benzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1938603

  SDS
alfa aesar™
A1938603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1938606

  SDS
alfa aesar™
A1938606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.15
Chemical Name or Material 4-(Trifluoromethyl)phenylacetonitrile
Assay 98%
Molecular Formula C9H6F3N
MDL Number MFCD00001921
CAS 2338-75-2
UN Number UN3439
Beilstein 909641
Synonym 4-(Trifluoromethyl)benzyl cyanide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 131° to 132°C (20mmHg)
Melting Point 43° to 47°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.