Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)phenylacetic acid, 99%

$684.07 - $684.07

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)phenylacetic acid
CAS 32857-62-8
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD00004352
Beilstein 2267687
Synonym (alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2252614

  SDS
alfa aesar™
B2252614
25g Each for $684.07
Description & Specifications

Specifications

Formula Weight 204.15
MDL Number MFCD00004352
Molecular Formula C9H7F3O2
Synonym (alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 79° to 81°C
Quantity 25g
CAS 32857-62-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Beilstein 2267687
Chemical Name or Material 4-(Trifluoromethyl)phenylacetic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.