Learn More
4-(Trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
$61.04 - $185.88
Chemical Identifiers
CAS | 1548-13-6 |
---|---|
Molecular Formula | C8H4F3NO |
Molecular Weight (g/mol) | 187.12 |
MDL Number | MFCD00002028 |
InChI Key | QZTWVDCKDWZCLV-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate |
PubChem CID | 73768 |
IUPAC Name | 1-isocyanato-4-(trifluoromethyl)benzene |
SMILES | FC(F)(F)C1=CC=C(C=C1)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1257603
|
Thermo Scientific Chemicals
L1257603 |
1 g |
Each for $61.04
|
|
|||||
AAL1257606
|
Thermo Scientific Chemicals
L1257606 |
5 g |
Each for $185.88
|
|
|||||
Description
4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
Chemical Identifiers
1548-13-6 | |
187.12 | |
QZTWVDCKDWZCLV-UHFFFAOYSA-N | |
73768 | |
FC(F)(F)C1=CC=C(C=C1)N=C=O |
C8H4F3NO | |
MFCD00002028 | |
4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate | |
1-isocyanato-4-(trifluoromethyl)benzene |
Specifications
1548-13-6 | |
91°C to 94°C (35 mmHg) | |
Pungent | |
1.471 | |
1 g | |
881041 | |
4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate | |
QZTWVDCKDWZCLV-UHFFFAOYSA-N | |
1-isocyanato-4-(trifluoromethyl)benzene | |
73768 | |
98% |
1.31 | |
70°C (158°F) | |
C8H4F3NO | |
MFCD00002028 | |
UN2285 | |
Moisture sensitive | |
It hydrolyzes in water. | |
FC(F)(F)C1=CC=C(C=C1)N=C=O | |
187.12 | |
187.12 | |
4-(Trifluoromethyl)phenyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H227-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P280-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H330-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 216-284-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only