missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)phenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1257603

 View more versions of this product

Catalog No. AAL1257603



Description

Description

Applications
4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
Specifications

Specifications

4-(Trifluoromethyl)phenyl isocyanate
Pungent
1g
C8H4F3NO
UN2285
4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate
C1=CC(=CC=C1C(F)(F)F)N=C=O
187.121
187.12
70°C (158°F)
91°C to 94°C (35mmHg)
1548-13-6
MFCD00002028
881041
QZTWVDCKDWZCLV-UHFFFAOYSA-N
1-isocyanato-4-(trifluoromethyl)benzene
73768
98%

Chemical Identifiers

1548-13-6
187.121
QZTWVDCKDWZCLV-UHFFFAOYSA-N
73768
C1=CC(=CC=C1C(F)(F)F)N=C=O
C8H4F3NO
MFCD00002028
4-trifluoromethyl phenyl isocyanate, 1-isocyanato-4-trifluoromethyl benzene, alpha,alpha,alpha-trifluoro-p-tolyl isocyanate, 4-trifluoromethyl phenylisocyanate, 4-isocyanatobenzotrifluoride, 4-trifluoromethylphenyl isocyanate, benzene, 1-isocyanato-4-trifluoromethyl, p-trifluoromethylphenyl isocyanate, isocyanic acid 4-trifluoromethyl phenyl ester, 4-trifluoromethyl benzenisocyanate
1-isocyanato-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H227-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATOBENZOTRIFLUORIDES

Recommended Storage : 2° to 8°C

Documents