4-(Trifluoromethyl)nicotinic acid, 97%, Maybridge
Manufacturer:  Fisher Scientific Company RF03948CB
 View more versions of this product
Catalog No. RF03948CB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
4-(Trifluoromethyl)nicotinic acid
250mg
97.0
C7H4F3NO2
3-carboxy-4-trifluoromethyl pyridine
C1=CN=CC(=C1C(F)(F)F)C(=O)O
191.109
191.11
Amber Glass Bottle
158063-66-2
97%
MFCD00082626
LMRJHNFECNKDKH-UHFFFAOYSA-N
4-(trifluoromethyl)pyridine-3-carboxylic acid
2777549
97%
Chemical Identifiers
158063-66-2
191.109
LMRJHNFECNKDKH-UHFFFAOYSA-N
2777549
C1=CN=CC(=C1C(F)(F)F)C(=O)O
C7H4F3NO2
MFCD00082626
3-carboxy-4-trifluoromethyl pyridine
4-(trifluoromethyl)pyridine-3-carboxylic acid
Safety and Handling
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS Documents