missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Trifluoromethyl)mandelic acid, 98%, Thermo Scientific Chemicals
$55.27 - $180.58
Chemical Identifiers
CAS | 395-35-7 |
---|---|
Molecular Formula | C9H7F3O3 |
Molecular Weight (g/mol) | 220.147 |
MDL Number | MFCD00009724 |
InChI Key | SDGXYUQKJPFLDG-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid |
PubChem CID | 164593 |
IUPAC Name | 2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid |
SMILES | C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2226403
|
Thermo Scientific Chemicals
B2226403 |
1 g |
Each for $55.27
|
|
|||||
AAB2226406
|
Thermo Scientific Chemicals
B2226406 |
5 g |
Each for $180.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
395-35-7 | |
220.147 | |
SDGXYUQKJPFLDG-UHFFFAOYSA-N | |
164593 | |
C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F |
C9H7F3O3 | |
MFCD00009724 | |
4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid | |
2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid |
Specifications
395-35-7 | |
C9H7F3O3 | |
1 g | |
4-trifluoromethyl mandelic acid, 2-hydroxy-2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylmandelic acid, hydroxy 4-trifluoromethyl phenyl acetic acid, 4-trifluoromethyl phenylglycolic acid, 2-4-trifluoromethyl phenyl-2-hydroxyacetic acid, p-trifluoromethylmandelic acid | |
C1=CC(=CC=C1C(C(=O)O)O)C(F)(F)F | |
220.147 | |
220.15 | |
4-(Trifluoromethyl)mandelic acid |
129°C to 131°C | |
MFCD00009724 | |
2979949 | |
SDGXYUQKJPFLDG-UHFFFAOYSA-N | |
2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid | |
164593 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-900-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only