Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)diphenyl ether, 97%

$174.78 - $174.78

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)diphenyl ether
CAS 2367-02-4
Molecular Formula C13H9F3O
Formula Weight 238.21
MDL Number MFCD01631641
Beilstein 1974851
Synonym 4-Phenoxybenzotrifluoride; 4-(Trifluoromethyl)phenyl phenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2411406

  SDS
alfa aesar™
B2411406
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 238.21
Molecular Formula C13H9F3O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Boiling Point 180°C (10mmHg)
Synonym 4-Phenoxybenzotrifluoride; 4-(Trifluoromethyl)phenyl phenyl ether
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2367-02-4
Quantity 5g
MDL Number MFCD01631641
Chemical Name or Material 4-(Trifluoromethyl)diphenyl ether
Beilstein 1974851
Assay 97%

Applications
4-(Trifluoromethyl)diphenyl ether is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.