Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Trifluoromethyl)cyclohexanecarboxylic acid, cis + trans, 97%

$42.04 - $388.74

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)cyclohexanecarboxylic acid
CAS 95233-30-0
Name Note Cis + Trans
Molecular Formula C8H11F3O2
Formula Weight 196.18
MDL Number MFCD00220049
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2728303

  SDS
alfa aesar™
H2728303
1g Each for $42.04

AAH2728314

  SDS
alfa aesar™
H2728314
25g Each for $388.74

AAH2728306

  SDS
alfa aesar™
H2728306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-(Trifluoromethyl)cyclohexanecarboxylic acid
Melting Point 88° to 94°C
Quantity 1g
Name Note Cis + Trans
CAS 95233-30-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C8H11F3O2
Formula Weight 196.18
MDL Number MFCD00220049
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.