Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)cyclohexan-1-one, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10356DE

 View more versions of this product

Catalog No. BTB10356DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 166.14
Quantity 5g
Assay 97%
Chemical Name or Material 4-(Trifluoromethyl)cyclohexan-1-one
Assay Percent Range 97%
Molecular Formula C7H9F3O
Linear Formula CF3C6H9(=O)
CAS 75091-99-5
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Trifluoromethyl)cyclohexan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9F3O
Linear Formula CF3C6H9(=O)
Formula Weight 166.14