Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)cyclohexan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)cyclohexan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9F3O
Linear Formula CF3C6H9(=O)
Formula Weight 166.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10356DE

 
fisher scientific company
BTB10356DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB10356EA

 
fisher scientific company
BTB10356EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.14
Quantity 5g
Assay 97%
Chemical Name or Material 4-(Trifluoromethyl)cyclohexan-1-one
Assay Percent Range 97%
Molecular Formula C7H9F3O
Linear Formula CF3C6H9(=O)
CAS 75091-99-5
Packaging Amber Glass Bottle