Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  434280010

 View more versions of this product

Catalog No. AC434280010

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 78° to 83°C
Color White
Quantity 1g
Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
CAS 683242-93-5
CAS Min % 94.0
CAS Max % 100.0
Physical Form White crystalline powder
Assay 94.0 % min. (GC)
Assay Percent Range 95%
Molecular Formula C13H20BF3O2
Formula Weight 276.11g/mol
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
CAS 683242-93-5
CAS Min % 94.0
CAS Max % 100.0
Assay 94.0 % min. (GC)
Assay Percent Range 95%
Molecular Formula C13H20BF3O2
Formula Weight 276.11g/mol
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents