Please login to your online account to display your discounted pricing

4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester, 95%, ACROS Organics™

$384.12 - $384.12

Chemical Identifiers

Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
CAS 683242-93-5
CAS Min % 94.0
CAS Max % 100.0
Assay 94.0 % min. (GC)
Assay Percent Range 95%
Molecular Formula C13H20BF3O2
Formula Weight 276.11g/mol
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC434280010

  SDS
thermo fisher scientific
434280010
Glass bottle 1g Each for $384.12

AC434280050

  SDS
thermo fisher scientific
434280050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 78° to 83°C
Color White
Quantity 1g
Chemical Name or Material 4-(Trifluoromethyl)cyclohex-1-enylboronic acid pinacol ester
CAS 683242-93-5
CAS Min % 94.0
CAS Max % 100.0
Physical Form White crystalline powder
Assay 94.0 % min. (GC)
Assay Percent Range 95%
Molecular Formula C13H20BF3O2
Formula Weight 276.11g/mol
Synonym 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)-1-cyclohexen-1-yl]-1,3,2-dioxaborolane
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.